siRNA Details
| Gene Symbol: Abcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008991.2 | NP_033017.2 | 9 | 828 | ||
| GenBank mRNA | AK004660.1 | - | 9 | 809 | ||
| AK144060.1 | - | 9 | 810 | |||
| AK153298.1 | - | 9 | 786 | |||
| BC009119.1 | - | 9 | 820 | 3154529 | ||
| BC054446.1 | - | 9 | 776 | 6477711 | ||
| CB245413.1 | - | 4 | 276 | |||

Abcd3
Genomic Map