siRNA Details
| Gene Symbol: Abcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008991.2 | NP_033017.2 | 23 | 2018 | ||
| GenBank mRNA | AK004660.1 | - | 23 | 1999 | ||
| AK144060.1 | - | 23 | 2000 | |||
| AK153298.1 | - | 23 | 1976 | |||
| BC009119.1 | - | 23 | 2010 | 3154529 | ||
| BC054446.1 | - | 23 | 1966 | 6477711 | ||
| X89569.1 | - | 14 | 1111 | |||

Abcd3
Genomic Map