siRNA Details
| Gene Symbol: Trpv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001445.2 | NP_001001445.1 | 13 | 2135 | ||
| GenBank mRNA | AB180097.1 | - | 12 | 1883 | ||
| AJ620495.1 | - | 12 | 1923 | |||
| AK051315.1 | - | 13 | 1993 | |||
| AY445519.1 | - | 12 | 1883 | |||
| AY452083.1 | - | 12 | 1916 | |||
| AY452084.1 | - | 12 | 1886 | |||

Trpv1
Genomic Map