siRNA Details
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277330.1 | NP_001264259.1 | 11 | 1160 | ||
| NM_001277805.1 | NP_001264734.1 | 12 | 1269 | |||
| NM_008995.2 | NP_033021.2 | 12 | 1232 | |||
| NM_175933.2 | NP_787947.1 | 11 | 1158 | |||
| GenBank mRNA | AJ416473.1 | - | 11 | 1140 | ||
| AK088886.1 | - | 11 | 1162 | |||
| AK145111.1 | - | 12 | 1205 | |||
| AK145361.1 | - | 11 | 1095 | |||
| AK161470.1 | - | 11 | 1155 | |||
| BC029748.1 | - | 12 | 1239 | 2599159 | ||
| Z97018.1 | - | 11 | 1157 | |||

Pex5
Genomic Map