siRNA Details
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277330.1 | NP_001264259.1 | 15 | 1831 | ||
| NM_001277805.1 | NP_001264734.1 | 16 | 1940 | |||
| NM_008995.2 | NP_033021.2 | 16 | 1903 | |||
| NM_175933.2 | NP_787947.1 | 15 | 1829 | |||
| GenBank mRNA | AJ416473.1 | - | 15 | 1811 | ||
| AK088886.1 | - | 15 | 1833 | |||
| AK145361.1 | - | 15 | 1766 | |||
| AK161470.1 | - | 15 | 1826 | |||
| BC029748.1 | - | 16 | 1910 | 2599159 | ||
| CF724027.1 | - | 4 | 485 | |||
| Z97018.1 | - | 15 | 1828 | |||

Pex5
Genomic Map