siRNA Details
| Gene Symbol: Pygm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011224.1 | NP_035354.1 | 18, 19 | 2370 | ||
| GenBank mRNA | AF124787.1 | - | 18, 19 | 2370 | ||
| AK019077.1 | - | 1, 2 | 149 | |||
| BC012961.1 | - | 18, 19 | 2376 | 3989941 | ||

Pygm
Genomic Map