siRNA Details
| Gene Symbol: Pygm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011224.1 | NP_035354.1 | 18 | 2290 | ||
| GenBank mRNA | AF124787.1 | - | 18 | 2290 | ||
| AK019077.1 | - | 1 | 69 | |||
| BC012961.1 | - | 18 | 2296 | 3989941 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pygm| Gene Symbol: Pygm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011224.1 | NP_035354.1 | 18 | 2290 | ||
| GenBank mRNA | AF124787.1 | - | 18 | 2290 | ||
| AK019077.1 | - | 1 | 69 | |||
| BC012961.1 | - | 18 | 2296 | 3989941 | ||