siRNA Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148673.3 | NP_683514.2 | 4, 5 | 503 | ||
| NM_198936.1 | NP_945174.1 | 3, 4 | 424 | |||
| GenBank mRNA | AK029117.1 | - | 3, 4 | 394 | ||
| AK046262.1 | - | 3, 4 | 395 | |||
| AK047597.1 | - | 4, 5 | 491 | |||
| AK049178.1 | - | 3, 4 | 393 | |||
| BC013810.1 | - | 23 | ||||
| BC025870.1 | - | 4, 5 | 482 | |||
| BC060954.1 | - | 3, 4 | 392 | |||
| BC082780.1 | - | 4, 5 | 483 | |||
| BC106099.1 | - | 3, 4 | 405 | |||
| BY243619.1 | - | 3, 4 | 428 | |||

Slu7
Genomic Map