siRNA Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148673.3 | NP_683514.2 | 14 | 1517 | ||
| NM_198936.1 | NP_945174.1 | 13 | 1438 | |||
| GenBank mRNA | AK029117.1 | - | 13 | 1408 | ||
| AK046262.1 | - | 13 | 1409 | |||
| AK047597.1 | - | 14 | 1505 | |||
| AK049178.1 | - | 13 | 1407 | |||
| BC013810.1 | - | 11 | 1037 | |||
| BC023057.1 | - | 3 | 292 | |||
| BC060954.1 | - | 13 | 1406 | |||
| BC082780.1 | - | 14 | 1497 | |||
| BC106099.1 | - | 13 | 1419 | |||

Slu7
Genomic Map