siRNA Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148673.3 | NP_683514.2 | 6 | 605 | ||
| NM_198936.1 | NP_945174.1 | 5 | 526 | |||
| GenBank mRNA | AK029117.1 | - | 5 | 496 | ||
| AK046262.1 | - | 5 | 497 | |||
| AK047597.1 | - | 6 | 593 | |||
| AK049178.1 | - | 5 | 495 | |||
| BC013810.1 | - | 3 | 125 | |||
| BC025870.1 | - | 6 | 584 | |||
| BC060954.1 | - | 5 | 494 | |||
| BC082780.1 | - | 6 | 585 | |||
| BC106099.1 | - | 5 | 507 | |||

Slu7
Genomic Map