siRNA Details
| Gene Symbol: Rab23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159729.1 | NP_001153201.1 | 5 | 651 | ||
| NM_008999.4 | NP_033025.2 | 5 | 763 | |||
| GenBank mRNA | AB232618.1 | - | 3 | 328 | ||
| AK016493.1 | - | 5 | 663 | |||
| AK019666.1 | - | 2 | 212 | |||
| AK045454.1 | - | 5 | 522 | |||
| AK047068.1 | - | 5 | 764 | |||
| AK082017.1 | - | 5 | 727 | |||
| AK164057.1 | - | 5 | 847 | |||
| BC025578.1 | - | 5 | 658 | 5323566 | ||
| Z22821.1 | - | 3 | 348 | |||

Rab23
Genomic Map