siRNA Details
| Gene Symbol: Rab33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016858.2 | NP_058554.1 | 2 | 850 | ||
| GenBank mRNA | AB004665.1 | - | 2 | 800 | ||
| AB232629.1 | - | 2 | 484 | |||
| AK004974.1 | - | 2 | 586 | |||
| AK087967.1 | - | 2 | 590 | |||
| AK139821.1 | - | 2 | 836 | |||
| AK144706.1 | - | 2 | 589 | |||
| AK159674.1 | - | 2 | 585 | |||
| AK163783.1 | - | 2 | 837 | |||
| AK170483.1 | - | 2 | 479 | |||
| BC065076.1 | - | 2 | 850 | 6815992 | ||

Rab33b
Genomic Map