siRNA Details
| Gene Symbol: Rab4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009003.3 | NP_033029.2 | 6 | 645 | ||
| GenBank mRNA | AB232591.1 | - | 6 | 505 | ||
| AK136566.1 | - | 6 | 634 | |||
| AK145452.1 | - | 6 | 628 | |||
| AK161877.1 | - | 6 | 613 | |||
| BC138508.1 | - | 6 | 558 | 8861528 | ||
| D86563.1 | - | 6 | 619 | |||

Rab4a
Genomic Map