siRNA Details
| Gene Symbol: Dennd5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021494.1 | NP_067469.1 | 23 | 4044 | ||
| GenBank mRNA | AJ245569.1 | - | 22 | 3950 | ||
| AK048609.1 | - | 14 | 1698 | |||
| BC022119.1 | - | 19 | 2697 | 5067385 | ||
| BC060230.1 | - | 23 | 4044 | 6840562 | ||

Dennd5a
Genomic Map