siRNA Details
| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044371.2 | NP_001037836.1 | 6 | 748 | ||
| NM_001283011.1 | NP_001269940.1 | 7 | 953 | |||
| NM_011233.3 | NP_035363.2 | 6 | 876 | |||
| GenBank mRNA | AF085737.1 | - | 6 | 799 | ||
| AJ011923.1 | - | 5 | 649 | |||
| AK134277.1 | - | 6 | 703 | |||
| AK162016.1 | - | 6 | 735 | |||
| AK167358.1 | - | 7 | 896 | |||
| BC023939.1 | - | 6 | 753 | |||
| BC050267.1 | - | 3 | 388 | |||
| BC066855.1 | - | 6 | 866 | 30473984 | ||

Rad17
Genomic Map