siRNA Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009012.2 | NP_033038.2 | 24, 25 | 4010 | ||
| GenBank mRNA | AK132949.1 | - | 4, 5 | 850 | ||
| U66887.1 | - | 24, 25 | 3988 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad50| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009012.2 | NP_033038.2 | 24, 25 | 4010 | ||
| GenBank mRNA | AK132949.1 | - | 4, 5 | 850 | ||
| U66887.1 | - | 24, 25 | 3988 | |||