siRNA Details
| Gene Symbol: Rad51b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252562.1 | NP_001239491.1 | 5, 6 | 515 | ||
| NM_009014.3 | NP_033040.2 | 5, 6 | 533 | |||
| GenBank mRNA | AK015771.1 | - | 5, 6 | 515 | ||
| AK032913.1 | - | 5, 6 | 534 | |||
| U92068.1 | - | 4, 5 | 475 | |||

Rad51b
Genomic Map