siRNA Details
Gene Symbol: Rnf185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145355.5 | NP_663330.2 | 2 | 831 | ||
GenBank mRNA | AK034108.1 | - | 2 | 201 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145355.5 | NP_663330.2 | 2 | 831 | ||
GenBank mRNA | AK034108.1 | - | 2 | 201 |