siRNA Details
| Gene Symbol: Rag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009019.2 | NP_033045.2 | 2 | 2127 | ||
| GenBank mRNA | BC138342.1 | - | 2 | 2052 | 8861362 | |
| M29475.1 | - | 2 | 2096 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rag1| Gene Symbol: Rag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009019.2 | NP_033045.2 | 2 | 2127 | ||
| GenBank mRNA | BC138342.1 | - | 2 | 2052 | 8861362 | |
| M29475.1 | - | 2 | 2096 | |||