siRNA Details
| Gene Symbol: Aldh1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009022.4 | NP_033048.2 | 8 | 926 | ||
| GenBank mRNA | AK078553.1 | - | 8 | 867 | ||
| BC075704.1 | - | 8 | 893 | 30471325 | ||
| X99273.1 | - | 8 | 871 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh1a2| Gene Symbol: Aldh1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009022.4 | NP_033048.2 | 8 | 926 | ||
| GenBank mRNA | AK078553.1 | - | 8 | 867 | ||
| BC075704.1 | - | 8 | 893 | 30471325 | ||
| X99273.1 | - | 8 | 871 | |||