siRNA Details
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009022.4 | NP_033048.2 | 4, 5 | 550 | ||
GenBank mRNA | AK078553.1 | - | 4, 5 | 491 | ||
BC075704.1 | - | 4, 5 | 517 | 30471325 | ||
X99273.1 | - | 4, 5 | 495 |
