siRNA Details
Gene Symbol: Mtmr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181409.3 | NP_852074.2 | 13 | 1432 | ||
GenBank mRNA | AK132375.1 | - | 13 | 1399 | ||
BC051083.1 | - | 13 | 1365 | 6486043 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mtmr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181409.3 | NP_852074.2 | 13 | 1432 | ||
GenBank mRNA | AK132375.1 | - | 13 | 1399 | ||
BC051083.1 | - | 13 | 1365 | 6486043 |