siRNA Details
| Gene Symbol: Mical3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270475.1 | NP_001257404.1 | 3 | 662 | ||
| NM_153396.3 | NP_700445.2 | 3 | 628 | |||
| GenBank mRNA | AF536756.1 | - | 3 | 591 | ||
| AK048201.1 | - | 3 | 640 | |||
| AK122497.2 | - | 3 | 487 | |||
| AK132669.1 | - | 3 | 612 | |||
| BC138257.1 | - | 3 | 589 | 8861276 | ||
| BC145214.1 | - | 3 | 589 | |||

Mical3
Genomic Map