siRNA Details
| Gene Symbol: Mical3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153396.3 | NP_700445.2 | 18 | 2853 | ||
| GenBank mRNA | AF536756.1 | - | 18 | 2816 | ||
| AK132669.1 | - | 18 | 2837 | |||
| BC138257.1 | - | 18 | 2814 | 8861276 | ||
| BC145214.1 | - | 19 | 2900 | |||
| BX630380.1 | - | 1 | 61 | |||

Mical3
Genomic Map