siRNA Details
| Gene Symbol: Reps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290633.1 | NP_001277562.1 | 9, 10 | 1319 | ||
| NM_178256.4 | NP_839987.2 | 9, 10 | 1322 | |||
| GenBank mRNA | AK147432.1 | - | 6, 7 | 601 | ||
| AY263368.1 | - | 9, 10 | 1267 | |||
| BC139012.1 | - | 9, 10 | 1198 | 8862034 | ||
| BC145500.1 | - | 9, 10 | 1195 | 9054063 | ||

Reps2
Genomic Map