siRNA Details
| Gene Symbol: Klf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178357.3 | NP_848134.1 | 3 | 515 | ||
| GenBank mRNA | AJ459773.2 | - | 3 | 361 | ||
| AK036618.1 | - | 3 | 501 | |||
| AK038397.1 | - | 3 | 613 | |||
| AK050156.1 | - | 3 | 517 | |||
| AK054170.1 | - | 3 | 487 | |||
| AK082838.1 | - | 3 | 504 | |||
| AK084913.1 | - | 3 | 488 | |||
| AK085319.1 | - | 3 | 488 | |||
| AK089044.1 | - | 3 | 495 | |||
| AK161233.1 | - | 3 | 507 | |||
| BC060642.1 | - | 3 | 513 | 6850350 | ||

Klf11
Genomic Map