siRNA Details
| Gene Symbol: Trim40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033235.3 | NP_001028407.1 | 2 | 950 | ||
| NM_001172168.1 | NP_001165639.1 | 2 | 950 | |||
| GenBank mRNA | AK136498.1 | - | 2 | 870 | ||
| BC147336.1 | - | 1 | 641 | 9056401 | ||
| BC147337.1 | - | 1 | 641 | 9056402 | ||
| BC172037.1 | - | 1 | 641 | 9054323 | ||

Trim40
Genomic Map