siRNA Details
| Gene Symbol: Utp14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001981.3 | NP_001001981.2 | 3 | 1364 | ||
| NM_001136226.1 | NP_001129698.1 | 4 | 1371 | |||
| GenBank mRNA | AK029972.1 | - | 4 | 1373 | ||
| AY316161.1 | - | 3 | 1151 | |||
| BC039778.1 | - | 1 | 111 | |||
| BC141102.1 | - | 2 | 1056 | 9055714 | ||

Utp14b
Genomic Map