siRNA Details
| Gene Symbol: Skint3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102474.1 | NP_001095944.1 | 9 | 1281 | ||
| NM_177578.4 | NP_808246.1 | 7 | 1241 | |||
| GenBank mRNA | AK040383.1 | - | 7 | 1242 | ||
| EF494894.1 | - | 9 | 1281 | |||
| EF494895.1 | - | 7 | 1065 | |||
| EF494896.1 | - | 8 | 933 | |||

Skint3
Genomic Map