siRNA Details
Gene Symbol: Magea9-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC116352.1 | - | 1 | 152 | ||
BC116353.1 | - | 1 | 154 | 40099901 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Magea9-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC116352.1 | - | 1 | 152 | ||
BC116353.1 | - | 1 | 154 | 40099901 |