siRNA Details
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009029.2 | NP_033055.2 | 20 | 2103 | ||
GenBank mRNA | AK132875.1 | - | 9 | 849 | ||
AK141028.1 | - | 20 | 2135 | |||
AK146467.1 | - | 15 | 1434 | |||
BC096525.1 | - | 20 | 2103 | 6832808 | ||
DQ400415.1 | - | 20 | 1953 | |||
M26391.1 | - | 20 | 2051 |
