siRNA Details
| Gene Symbol: Rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009029.2 | NP_033055.2 | 17 | 1684 | ||
| GenBank mRNA | AK132875.1 | - | 6 | 430 | ||
| AK141028.1 | - | 17 | 1716 | |||
| AK146467.1 | - | 12 | 1015 | |||
| BC096525.1 | - | 17 | 1684 | 6832808 | ||
| DQ400415.1 | - | 17 | 1534 | |||
| M26391.1 | - | 17 | 1632 | |||

Rb1
Genomic Map