siRNA Details
| Gene Symbol: Rbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011249.2 | NP_035379.2 | 20 | 2847 | ||
| GenBank mRNA | AK156055.1 | - | 20 | 2826 | ||
| BC060124.1 | - | 20 | 2814 | 6408844 | ||
| BC069179.1 | - | 12 | 1625 | |||
| U27177.1 | - | 20 | 2831 | |||
| U33320.1 | - | 20 | 2814 | |||

Rbl1
Genomic Map