siRNA Details
| Gene Symbol: Rbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282000.1 | NP_001268929.1 | 2 | 437 | ||
| NM_001282001.1 | NP_001268930.1 | 2 | 437 | |||
| NM_011250.4 | NP_035380.3 | 2 | 437 | |||
| GenBank mRNA | AK142172.1 | - | 2 | 396 | ||
| AK155256.1 | - | 2 | 401 | |||
| AK160027.1 | - | 2 | 384 | |||
| BC020528.1 | - | 2 | 385 | 4240592 | ||
| BY749910.1 | - | 2 | 401 | |||
| U36799.1 | - | 2 | 337 | |||
| U47333.1 | - | 2 | 379 | |||
| U50850.1 | - | 2 | 345 | |||

Rbl2
Genomic Map