siRNA Details
| Gene Symbol: Rbm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011251.3 | NP_035381.2 | 4, 5 | 1656 | ||
| NM_029169.3 | NP_083445.1 | 4, 5 | 1927 | |||
| GenBank mRNA | AJ006486.2 | - | 4, 5 | 1535 | ||
| AK077899.1 | - | 4, 5 | 1834 | |||
| AK081846.1 | - | 4, 5 | 1584 | |||
| AK134164.1 | - | 4, 5 | 1567 | |||
| AK145612.1 | - | 4, 5 | 1816 | |||
| AK220446.1 | - | 4, 5 | 1805 | |||
| BC026129.1 | - | 4, 5 | 1681 | 3982861 | ||
| BC026130.1 | - | 4, 5 | 1830 | |||
| X96701.1 | - | 2, 3 | 311 | |||

Rbm6
Genomic Map