siRNA Details
| Gene Symbol: Rbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015745.2 | NP_056560.1 | 3 | 3422 | ||
| GenBank mRNA | BC017610.1 | - | 3 | 3422 | 4527589 | |
| BC023069.1 | - | 3 | 713 | |||
| BC034121.1 | - | 3 | 541 | |||
| BC035273.1 | - | 3 | 1854 | |||
| BC040282.1 | - | 3 | 3406 | |||
| BC042480.1 | - | 3 | 3384 | 5363924 | ||
| BC042509.1 | - | 3 | 615 | |||
| BC083133.1 | - | 3 | 3441 | 6492729 | ||

Rbp3
Genomic Map