siRNA Details
| Gene Symbol: Rbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015745.2 | NP_056560.1 | 1 | 2577 | ||
| GenBank mRNA | BC017610.1 | - | 1 | 2577 | 4527589 | |
| BC035273.1 | - | 1 | 1009 | |||
| BC040282.1 | - | 1 | 2561 | |||
| BC042480.1 | - | 1 | 2539 | 5363924 | ||
| BC083133.1 | - | 1 | 2596 | 6492729 | ||

Rbp3
Genomic Map