siRNA Details
| Gene Symbol: Rfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011258.2 | NP_035388.2 | 17 | 2391 | ||
| GenBank mRNA | AK134660.1 | - | 2 | 169 | ||
| BC085173.1 | - | 17 | 2368 | 30549691 | ||
| BC085174.1 | - | 17 | 2358 | |||
| U01222.1 | - | 17 | 2359 | |||
| U07157.1 | - | 17 | 2354 | |||
| U36441.1 | - | 17 | 2365 | |||
| X72711.1 | - | 17 | 2357 | |||

Rfc1
Genomic Map