siRNA Details
| Gene Symbol: Reln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011261.2 | NP_035391.2 | 44, 45 | 7224 | ||
| GenBank mRNA | D63520.1 | - | 3, 4 | 474 | ||
| U24703.1 | - | 44, 45 | 7209 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reln| Gene Symbol: Reln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011261.2 | NP_035391.2 | 44, 45 | 7224 | ||
| GenBank mRNA | D63520.1 | - | 3, 4 | 474 | ||
| U24703.1 | - | 44, 45 | 7209 | |||