siRNA Details
| Gene Symbol: Ret | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080780.1 | NP_001074249.1 | 5, 6 | 1676 | ||
| NM_009050.2 | NP_033076.2 | 5, 6 | 1676 | |||
| GenBank mRNA | AF209436.1 | - | 5, 6 | 1065 | ||
| AK044686.1 | - | 5, 6 | 1263 | |||
| AK051633.1 | - | 5, 6 | 1255 | |||
| AK084977.1 | - | 5, 6 | 1326 | |||
| AY326397.1 | - | 5, 6 | 1065 | |||
| BC059012.1 | - | 5, 6 | 1650 | 6849696 | ||
| X67812.1 | - | 5, 6 | 1065 | |||

Ret
Genomic Map