siRNA Details
| Gene Symbol: Ripk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009068.3 | NP_033094.3 | 9, 10 | 1677 | ||
| GenBank mRNA | AK030959.1 | - | 9, 10 | 1908 | ||
| AK156803.1 | - | 9, 10 | 1942 | |||
| BC050905.1 | - | 9, 10 | 1677 | 5721177 | ||
| BC054542.1 | - | 9, 10 | 1677 | 5721177 | ||
| BC058162.1 | - | 9, 10 | 1863 | 5135235 | ||
| U25995.1 | - | 8, 9 | 1592 | |||

Ripk1
Genomic Map