siRNA Details
| Gene Symbol: Rlim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011276.3 | NP_035406.3 | 5 | 1976 | ||
| GenBank mRNA | AK013207.1 | - | 5 | 1948 | ||
| AK029295.1 | - | 5 | 1970 | |||
| BC012960.1 | - | 3 | 1709 | 4193003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rlim| Gene Symbol: Rlim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011276.3 | NP_035406.3 | 5 | 1976 | ||
| GenBank mRNA | AK013207.1 | - | 5 | 1948 | ||
| AK029295.1 | - | 5 | 1970 | |||
| BC012960.1 | - | 3 | 1709 | 4193003 | ||