siRNA Details
| Gene Symbol: Trim10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011280.2 | NP_035410.2 | 6, 7 | 1225 | ||
| GenBank mRNA | AF134811.1 | - | 6, 7 | 1207 | ||
| AF220121.1 | - | 6, 7 | 1212 | |||
| AK011082.1 | - | 6, 7 | 1225 | |||
| BC051632.1 | - | 6, 7 | 1198 | 6507331 | ||

Trim10
Genomic Map