siRNA Details
| Gene Symbol: Robo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019413.2 | NP_062286.2 | 13, 14 | 2972 | ||
| GenBank mRNA | Y17793.1 | - | 13, 14 | 2234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Robo1| Gene Symbol: Robo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019413.2 | NP_062286.2 | 13, 14 | 2972 | ||
| GenBank mRNA | Y17793.1 | - | 13, 14 | 2234 | ||