siRNA Details
| Gene Symbol: Slc22a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164634.1 | NP_001158106.1 | 4 | 748 | ||
| NM_001164635.1 | NP_001158107.1 | 4 | 656 | |||
| NM_031194.5 | NP_112471.3 | 5 | 828 | |||
| GenBank mRNA | AB079895.1 | - | 3 | 485 | ||
| AF078869.1 | - | 4 | 603 | |||
| AK031962.1 | - | 5 | 794 | |||
| AK044336.1 | - | 4 | 625 | |||
| AK157891.1 | - | 4 | 657 | |||
| BC014762.1 | - | 4 | 634 | 4239544 | ||
| BU615016.1 | - | 4 | 698 | |||

Slc22a8
Genomic Map