siRNA Details
| Gene Symbol: Slc22a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164634.1 | NP_001158106.1 | 2 | 332 | ||
| NM_001164635.1 | NP_001158107.1 | 2 | 240 | |||
| NM_031194.5 | NP_112471.3 | 3 | 412 | |||
| GenBank mRNA | AB079895.1 | - | 1 | 69 | ||
| AF078869.1 | - | 2 | 187 | |||
| AK031962.1 | - | 3 | 378 | |||
| AK044336.1 | - | 2 | 209 | |||
| AK157891.1 | - | 2 | 241 | |||
| BB622154.1 | - | 3 | 375 | |||
| BB843926.1 | - | 2 | 241 | |||
| BC014762.1 | - | 2 | 218 | 4239544 | ||
| BU615016.1 | - | 2 | 282 | |||

Slc22a8
Genomic Map