siRNA Details
| Gene Symbol: Rpgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177950.1 | NP_001171421.1 | 18 | 3202 | ||
| NM_001177952.1 | NP_001171423.1 | 17 | 2317 | |||
| NM_011285.2 | NP_035415.1 | 17 | 2323 | |||
| GenBank mRNA | AF044677.1 | - | 17 | 2241 | ||
| AJ238396.1 | - | 18 | 3114 | |||
| AK136298.1 | - | 17 | 2223 | |||
| AK138977.1 | - | 17 | 2318 | |||

Rpgr
Genomic Map