siRNA Details
| Gene Symbol: Rpgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177950.1 | NP_001171421.1 | 10 | 1369 | ||
| NM_001177951.1 | NP_001171422.1 | 10 | 1369 | |||
| NM_001177952.1 | NP_001171423.1 | 10 | 1363 | |||
| NM_001177953.1 | NP_001171424.1 | 10 | 1369 | |||
| NM_011285.2 | NP_035415.1 | 10 | 1369 | |||
| GenBank mRNA | AF044677.1 | - | 10 | 1287 | ||
| AJ238396.1 | - | 10 | 1281 | |||
| AK017192.1 | - | 10 | 1301 | |||
| AK029864.1 | - | 10 | 1495 | |||
| AK038652.1 | - | 10 | 1333 | |||
| AK136298.1 | - | 10 | 1300 | |||
| AK138977.1 | - | 10 | 1364 | |||
| BC012661.1 | - | 10 | 1308 | |||

Rpgr
Genomic Map