siRNA Details
| Gene Symbol: Rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011302.3 | NP_035432.3 | 4, 5 | 507 | ||
| GenBank mRNA | AF073780.1 | - | 4, 5 | 370 | ||
| AF084561.1 | - | 4, 5 | 378 | |||
| AJ011381.1 | - | 4, 5 | 358 | |||
| BC046422.1 | - | 4, 5 | 507 | 5401012 | ||

Rs1
Genomic Map