siRNA Details
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003933.2 | NP_001003933.1 | 3 | 2507 | ||
NM_001003934.2 | NP_001003934.1 | 4 | 2564 | |||
NM_001271486.1 | NP_001258415.1 | 4 | 1601 | |||
NM_001271487.1 | NP_001258416.1 | 3 | 440 | |||
NM_053076.3 | NP_444306.1 | 2 | 383 | |||
GenBank mRNA | AB046114.1 | - | 2 | 377 | ||
AF195940.1 | - | 2 | 327 | |||
AK032109.1 | - | 4 | 1578 | |||
AK036892.1 | - | 2 | 349 | |||
AK049845.1 | - | 2 | 315 | |||
AK075883.1 | - | 2 | 312 | |||
AK088670.1 | - | 2 | 315 | |||
AK146505.1 | - | 2 | 191 | |||
AK148792.1 | - | 2 | 342 | |||
AK148947.1 | - | 2 | 344 | |||
AK165683.1 | - | 2 | 314 | |||
AK197640.1 | - | 1 | 33 | |||
AY164700.1 | - | 2 | 377 | |||
AY427822.1 | - | 3 | 2488 | |||
AY507126.1 | - | 2 | 175 | |||
AY507127.1 | - | 2 | 2110 | |||
AY750849.1 | - | 4 | 2545 | |||
BC005478.1 | - | 2 | 286 | |||
BC014697.1 | - | 2 | 294 | |||
BC036717.1 | - | 2 | 292 | |||
BC079882.1 | - | 4 | 2474 | 6839323 |
